Order Entry
ContactUsLinkComponent
VWR Boston Round Bottles
VWR® Laboratory Narrow Mouth Bottles, PP
Catalog #: 414004-182
Supplier:  VWR International
VWR Boston Round Bottles
VWR® Laboratory Narrow Mouth Bottles, PP
Catalog #: 414004-182
Supplier:  VWR International

Specifications

  • Cap Size:
    28-410
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    414004-182

Specifications

About this item

Specimen bottles are for general laboratory use, especially where sterilization is necessary.

  • Clear polypropylene construction
  • Screw enclosure included
  • Excellent chemical resistance
  • Can be stored in freezer to −40 °C (−40 °F)
  • Can be autoclaved with or without contents

Bottles are translucent and provide better clarity than polyethylene bottles

Note: Before autoclaving, set closure on top of container without engaging threads.

Delivery: Polypropylene screw closure included.
Customers Who Bought This Also Bought