Order Entry
ContactUsLinkComponent
VWR Laboratory Bottles
VWR® Laboratory Bottles, LDPE
Catalog #: 414004-174
Supplier:  VWR International
VWR Laboratory Bottles
VWR® Laboratory Bottles, LDPE
Catalog #: 414004-174
Supplier:  VWR International

Specifications

  • Cap Size:
    28-410
  • Capacity:
    500 ml (16.9 oz.)
  • Description:
    Narrow mouth bottles

Specifications

About this item

Translucent, flexible bottles are for general laboratory use, and are especially good for shipping liquids and for trace metal analysis.

  • Leakproof
  • Translucent siding

Can be stored in freezer to −100 °C (−148 °F). Offer better visibility of contents than high-density polyethylene bottles, and have excellent chemical resistance to most acids, bases, and alcohols.

Delivery: Polypropylene screw closures included.
Customers Who Bought This Also Bought