Order Entry
ContactUsLinkComponent
VWR Boston Round Bottles
VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
Catalog #: 414004-166
Supplier:  VWR International
VWR Boston Round Bottles
VWR® Narrow Mouth Laboratory Bottles, Amber HDPE
Catalog #: 414004-166
Supplier:  VWR International

Specifications

  • Cap Size:
    24-410
  • Capacity:
    250 ml (8.5 oz.)
  • Cat. no.:
    414004-166

Specifications

About this item

Bottles are for general laboratory use and for storing, shipping, and packing liquids, especially light-sensitive materials or photosensitive samples.

  • Amber coloring protects light-sensitive contents
  • High-density polyethylene fabrication
  • Excellent chemical resistance

Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to most acids, bases, and alcohols. Amber polypropylene screw closure included. Bottles comply with current USP requirements for maximum light transmission.

Customers Who Bought This Also Bought