Order Entry
ContactUsLinkComponent
VWR Boston Round Bottles
VWR® Laboratory Bottles, HDPE
Catalog #: 414004-155
Supplier:  VWR International
VWR Boston Round Bottles
VWR® Laboratory Bottles, HDPE
Catalog #: 414004-155
Supplier:  VWR International

Specifications

  • Cap Size:
    20-415
  • Capacity:
    60 ml (2 oz.)
  • Description:
    Narrow mouth bottles
  • Cat. no.:
    414004-155

Specifications

About this item

Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.

  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding

Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives.

Delivery: Polypropylene screw closure included.
Customers Who Bought This Also Bought