VWR® Disposable Transfer Pipettes
Catalog #: 414004-004
Supplier: VWR International
Specifications
- Capacity:7.5 ml
- Sterility:Non sterile
- Description:Standard
- Cat. no.:414004-004
- Packaging:Bulk packaged
- Overall Length:15.5 cm (6¹/₈")
- Bulb Draw:3.2 ml
- Drops per mL:20
Specifications
About this item
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
- Will not shatter or break
- Can be used in liquid nitrogen
- Nontoxic and inert
- No bulb to insert or remove
- Uniform drop size
Molded from see-through low density polyethylene, they are inert to biological fluids and most acids. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Transfer pipettes can be sealed and refrigerated, and will work well whenever there is a need for quick, safe fluid transfer. Can be gas sterilized.
Customers Who Bought This Also Bought