Order Entry
ContactUsLinkComponent
GR515 Fluted Filter Paper Circles
Pleated (Fluted) Filter Paper, Grade 515, Ahlstrom
Catalog #: 38000-200
Supplier:  Ahlstrom
GR515 Fluted Filter Paper Circles
Pleated (Fluted) Filter Paper, Grade 515, Ahlstrom
Catalog #: 38000-200
Supplier:  Ahlstrom

Specifications

  • Ø:
    18.5 cm
  • Flow rate:
    Fast
  • Retention:
    25 µm
  • Load capacity:
    High
  • Cat. no.:
    38000-200
  • Supplier No.:
    5150-1850

Specifications

About this item

Ahlstrom Grade 515 is the pre-pleated version of Grade 615.

Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.

Customers Who Bought This Also Bought