Order Entry
Canada
ContactUsLinkComponent
NALGENE* Pipet Jar
Nalgene® Pipette Jars, Thermo Scientific
Catalog # 36322-100
undefined
Nalgene® Pipette Jars, Thermo Scientific
Catalog # 36322-100
Supplier Number:  5242-0050

Specifications

  • Dimensions:
    16.5Dia. x 68.6H cm (61/2 x 27")
  • Cat. no.:
    36322-100
  • For:
    Pipettes up to 40.6 cm (16") length
  • Supplier No.:
    5242-0050

Specifications

About this item

Cross-linked high-density polyethylene pipette jars are ideal for soaking and washing pipettes and glassware, and as large general-purpose containers

Caution: Concentrated sulfuric acid will cause jar to fail. When using sulfuric acid dichromate compound, the acid concentration must be 60% or less. See label on jar. Do not mix sulfuric acid dichromate compound in jar. Keep jar on drain board, in sink, or in container large enough to contain contents in case of leakage.

Tough, stiff, seamless jars resist stress-cracking. All sizes have flanged base for added stability. Breakage is sharply reduced and accidents avoided.

Ordering information: Supplied with dust covers. Place pipettes in the corresponding Nalgene pipette basket (53596-021 series; sold separately) for easy submersion in and retrieval from the jar.

Baskets

Product Information