Order Entry
ContactUsLinkComponent
BUFFER TRI-CHEK CAPSULE SET
Certified pH Buffer Set
Catalog #: 34175-485
Supplier:  Micro Essential
CAS Number:  
BUFFER TRI-CHEK CAPSULE SET
Certified pH Buffer Set
Catalog #: 34175-485
Supplier:  Micro Essential
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Kit, Certified pH Buffer Set, GLOBE Approved Product
  • Supplier No.:
    CC-471V

Specifications

About this item

Set includes calibrated and pre-weighed amounts of buffer powders at three pH values (pH 4.0, 7.0, and 10.0) for testing or calibrating your pH meter.

Powders come in capsules and each is sufficient for 100 ml of buffer solution.

Delivery: Includes 10 capsules for each pH value and a vial of buffer preservative.
Customers Who Bought This Also Bought