VWR® Tubular Knit Mop Heads
Catalog #: 33502-728
Supplier: VWR International
Small Business Enterprise
Specifications
- Description:Tubular knit mop head
Specifications
About this item
Mop heads are designed to have excellent sorbency and are suitable for use in cleanroom conditions.
- Tubular-knit, 100% polyester string mop heads
- Excellent sorbency
- Mop heads are double clear poly-bagged for cleanroom use
Mop head 33502-730 is sterile. Mop head 33502-732 features ribbing. Appropriate for ISO Class 4 (FED-STD-209E Class 10/M2.5) conditions.
Customers Who Bought This Also Bought