Polyethylene [ST] Stoppers, Chemglass
Catalog #: 33501-434
Supplier: Chemglass
Small Business Enterprise
Specifications
- ST Joint No.:24/25
- Cat. no.:33501-434
- Supplier No.:CG-3021-03
Specifications
About this item
Inexpensive, tapered stoppers provide a tight, leakproof fit while the top ring permits the stopper to be removed easily
Stoppers are made of yellow polyethylene.
Customers Who Bought This Also Bought