Nalgene® Separatory Funnels, Polypropylene, Squibb, Pear-Shape, Thermo Scientific
Catalog #: 30356-802
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:53 mm (23/32")
- Capacity:1000 ml
- Cat. no.:30356-802
- Supplier No.:4300-1000
Specifications
About this item
Translucent, leakproof funnels feature excellent chemical resistance and do not require lubrication
Compatible with a wide range of lab reagents. Separation lines between immiscible liquids are easy to observe. Funnel and screw closure are autoclavable; stopcock assembly may be chemically disinfected.
Customers Who Bought This Also Bought