KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Catalog #: 30353-054
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:500 ml
- Cat. no.:30353-054
- Stopper No.:27
- Handle Color:Red
- Stopcock Size:4
- Supplier No.:29052F-500
Specifications
About this item
Designed with large lower stem I.D
for more complete separation. RAY-SORB® processed to protect materials sensitive to light of shorter wavelengths. Color-coded stopper. Designed from ASTM Specification E1096, Type IV requirements.
Customers Who Bought This Also Bought