Order Entry
ContactUsLinkComponent
Filter Paper Discs for Buchner Funnels, Polyethylene, Table-Top
SP Bel-Art Filter Paper Discs for Buchner Table-Top Funnels, Bel-Art Products, a part of SP
Catalog #: 30309-002
SP Bel-Art Filter Paper Discs for Buchner Table-Top Funnels, Bel-Art Products, a part of SP
Catalog #: 30309-002

Specifications

  • Description:
    Cellulose filter paper discs for 45.7 cm (18") I.D. funnels
  • Cat. no.:
    30309-002
  • Supplier No.:
    146320018

Specifications

About this item

These heavy bond grade filter paper discs are custom-sized to fit table-top Buchner funnels and have a thickness of 0.30 mm.

  • Made from creped white cellulose to improve porosity
  • Flow rate: 90 ml per minute
  • Wet burst strength is greater than 68 KPa; able to retain particles as small as 3 microns
Customers Who Bought This Also Bought