SP Bel-Art 'Wire' Lead Flask Weight with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Catalog #: 29700-066
Supplier: Bel-Art Products, a Part of SP
Specifications
- Cat. no.:29700-066
- Length:30 cm (12")
- Weight:113 g (4 oz.)
- Supplier No.:183240000
Specifications
About this item
These lead weights can be coiled into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath.
- Red vinyl coating
- Available in two different weights
- Chemical-resistant
- Flexible and versatile
They can be coiled around flasks, threaded through handles or looped around bottle necks.
Available in two weights, they are coated with chemical resistant red Vikem® vinyl. The 30 cm (12") length and flexibility make them very versatile.
Customers Who Bought This Also Bought