PYREX® Snap Caps, Polyethylene, Corning
Catalog #: 29610-579
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Cap Size:500
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:29610-579
- Supplier No.:7666-500
Specifications
About this item
These blue caps provide an economical closure for certain volumetric flasks (see 29610-004 and -285 series).
Customers Who Bought This Also Bought