Corning® 96-well Clear Polystyrene Microplates, Corning
Catalog #: 29442-056
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Well Volume:360 µl
- Color:Clear
- Environmentally Preferable:Low Manufacturing Impact
- Material:Polystyrene
- No. of wells:96
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:29442-056
- Packaging:Individually wrapped
- Surface Treatment:Tissue culture (TC)
- Bottom Style:Flat-Bottom
- Supplier No.:3596
Specifications
About this item
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
NA Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. NA Cat. Nos. 3585 and 3595 have special low evaporation lids. NA Cat. No. 3841 is aseptically manufactured.
Customers Who Bought This Also Bought