Order Entry
ContactUsLinkComponent
NALGENE* Analytical Test Filter Funnels, Sterile
Nalgene® Analytical Test Filter Funnels, Sterile, Thermo Scientific
Catalog #: 28198-865
NALGENE* Analytical Test Filter Funnels, Sterile
Nalgene® Analytical Test Filter Funnels, Sterile, Thermo Scientific
Catalog #: 28198-865

Specifications

  • Pore size:
    0.45 µm
  • Volume:
    250 ml
  • Description:
    Filter funnel with membrane grid
  • Supplier No.:
    145-2045

Specifications

About this item

For microbiological testing of water quality, food/beverages, raw material, sterility, and finished product quality.

Each disposable, sterile filter funnel consists of a polypropylene upper chamber, a high-impact polystyrene collar, and a cellulose nitrate membrane. Graduated upper chamber separates easily from the collar for easy access to the membrane. Funnel adapters allow use with No. 8 stoppers; six adapters are included with each case. The 0.45µm pore size membranes are certified for water quality work including coliform analysis. Membrane diameter: 47mm.

  • Membrane Ø: 47 mm

For microbiological testing of water quality, food/beverages, raw material, sterility, and finished product quality.

Each disposable, sterile filter funnel consists of a polypropylene upper chamber, a high-impact polystyrene collar, and a cellulose nitrate membrane. Graduated upper chamber separates easily from the collar for easy access to the membrane. Funnel adapters allow use with No. 8 stoppers; six adapters are included with each case. The 0.45 µm pore size membranes are certified for water quality work including coliform analysis.

Each disposable, sterile filter funnel consists of a polypropylene upper chamber, a high-impact polystyrene collar, and a cellulose nitrate membrane. Graduated upper chamber separates easily from the collar for easy access to the membrane. Funnel adapters allow use with No. 8 stoppers; six adapters are included with each case. The 0.45 µm pore size membranes are certified for water quality work including coliform analysis.

Caution: For lab use only. Not for in-vitro diagnosis or parenterals.
Customers Who Bought This Also Bought