Order Entry
ContactUsLinkComponent
DIALYSIS MEMBRANES 23MM RL100
Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
Catalog #: 28170-138
Small Business Enterprise
DIALYSIS MEMBRANES 23MM RL100
Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
Catalog #: 28170-138
Small Business Enterprise

Specifications

  • Ø:
    14.6 mm
  • Length:
    30.5 m
  • Flat Width:
    23 mm
  • Supplier No.:
    S632650

Specifications

About this item

Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.

The clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications. Possessing a broad range of chemical compatibilities, Standard RC membranes can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols, and some mild or dilute organic solvents, including DMSO.

Customers Who Bought This Also Bought