Barnstead Replacement Cartridges, Thermo Scientific
Catalog #: 26303-256
Supplier: Thermo Fisher Scientific
Specifications
- Description:Ultrapure, Mixed-Bed Resin
- Cat. no.:26303-256
- For Use With:NANOpure® II, Older Style NANOpure® Systems, B-Pure™, Bantam, Cooling Loops
- Capacity:760 grains as NaCl to a 1 Megohm Endpoint
- Supplier No.:D0809
Specifications
About this item
Cartridges feature all polypropylene construction and are designed to fit certain NANOpure®, E-pure™, B-pure™, PCS, Bantam holder, or cooling loop systems.
Customers Who Bought This Also Bought