Coplin Staining Jar with Cover, Wheaton
Catalog #: 25460-000
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Specifications
- Dimensions:3 I.D. x 7.6 H cm (13/16 x 3")
- Supplier No.:900470
Specifications
About this item
Heavy soda lime glass, with ground-on cover and wide, flat base, for convenient manipulation of sections attached to slides
Holds ten 75x25mm (3x1") slides.
Customers Who Bought This Also Bought