Order Entry
ContactUsLinkComponent
VWR* Disposable Aluminum Weighing Dishes
VWR® Disposable Aluminum Weighing Dishes
Catalog #: 25433-020
Supplier:  VWR International
VWR* Disposable Aluminum Weighing Dishes
VWR® Disposable Aluminum Weighing Dishes
Catalog #: 25433-020
Supplier:  VWR International

Specifications

  • Capacity:
    75 ml
  • Cat. no.:
    25433-020
  • I.D.×D:
    6.5×3.0 cm (2⁹⁄₁₆×1³⁄₁₆")

Specifications

About this item

Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.

  • Made of light-gauge aluminum
  • Stackable
  • Contaminant-free
  • Petroleum-free

Pans feature a flanged edge with curled lip and are contaminate- and petroleum-free.

Customers Who Bought This Also Bought