VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Catalog #: 25384-152
Supplier: VWR International
Specifications
- Capacity:100 ml (3.4 oz.)
- Cat. no.:25384-152
Specifications
About this item
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
- Polypropylene construction
- Transparent
- Graduated markings
- Disposable design
Graduated, with accuracy of ±5%. May also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Write-on cardboard caps ensure spillproof, leak-resistant closure.
Customers Who Bought This Also Bought