DESICCATOR SCHEIBLER 250MM
Catalog #: 25012-065
Supplier: Variety Glass
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61
Hydrogen peroxide 35% (w/w) in aqueous solution stabilized
Supplier: Thermo Scientific Chemicals
$311.01
1-Heptanesulfonic acid sodium salt monohydrate ≥99% for HPLC, for ion pair chromatography, Sigma-Aldrich®
Supplier: MilliporeSigma
$1,619.66