SP Bel-Art Desi Can™ Reusable Desiccant Canister, Bel-Art Products, a part of SP
Catalog #: 24988-350
Supplier: Bel-Art Products, a Part of SP
Specifications
- Cat. no.:24988-350
- L×W×H:5.1×10.2×1.3 cm (2×4×¹/₂")
- Supplier No.:420460000
Specifications
About this item
Silica gel effectively absorbs moisture through perforations in the aluminum case.
- Reusable design
- Monitor changes color with saturation level
- Unlimited potential life
One canister of Reusable Desi-Can™ Canister maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.) of space.
Excellent for use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture.
It can be regenerated in a 150º C (300º F) oven in about 3 hours for unlimited life. A monitor window shows pink when recharged and green when saturated.
Customers Who Bought This Also Bought