Nalgene® CryoCane™ Holders, Aluminum, Thermo Scientific
Catalog #: 24270-029
Supplier: Thermo Fisher Scientific
Specifications
- Color:White
- Description:CryoCane™ coders
- Cat. no.:24270-029
- Supplier No.:DS5020-0000
Specifications
About this item
Firmly holds NALGENE® cryogenic vials for storage in Dewar-type, liquid-nitrogen freezers.
After cooling vials to storage temperature, snap them into the cane end-to-end with vial marking area facing out. Cane projects inward slightly to support base of each vial. Accommodates most other brands of cryogenic vials. Identify canes with CryoCane™ color-coded tabs.
Unique, colorful tabs slip onto the end of a cane to identify a particular batch of vials or a specific technician’s work.
Remain firmly in place during storage in mechanical or liquid-nitrogen freezers. Flat writing surface for additional identification.
Customers Who Bought This Also Bought