Order Entry
ContactUsLinkComponent
VWR* Robo* Thermometer
VWR® Traceable® Robo™ Thermometers
Catalog #: 23609-206
Supplier:  VWR International
VWR* Robo* Thermometer
VWR® Traceable® Robo™ Thermometers
Catalog #: 23609-206
Supplier:  VWR International

Specifications

  • Probe:
    20.3L x 0.36 dia. cm (8 x 1/8")
  • Resolution:
    0.1° from –20° to 200°, 1° elsewhere
  • Description:
    Robo™ Thermometer
  • Temperature range:
    –50° to 280°C (–58° to 536°F)
  • Accuracy:
    ±1.0 °C between –20 and 100 °C
  • Length:
    23.5 cm (91/4")

Specifications

About this item

Ideal for reading temperatures in reagents, water baths, foods, air, and semisolids.

Ideal for reading temperatures in reagents, water baths, foods, air, and semisolids. Great for use in any deep vessel. Durable stainless steel and plastic construction makes these thermometers safer than mercury thermometers. Digital display conveniently rotates 180° to a vertical position, or to a horizontal position, for viewing sample and temperature simultaneously. Bright LCD digits are 6.4mm (1/4") high and easier to read than pointer or mercury thermometers.

  • Traceable®
  • Stainless steel probe
  • Rotating digital display
  • Read °C and °F
  • Extra-wide range

Great for use in any deep vessel

Great for use in any deep vessel

Probe is resistant to acids, bases, solvents, and most laboratory chemicals. Long stem allows user to make measurements while stirring solutions. Change between °F and °C with the press of a button. Readings are updated every second. Weigh less than 28g (1 oz.).

Durable stainless steel and plastic construction makes these thermometers safer than mercury thermometers. Digital display conveniently rotates 180° to a vertical position, or to a horizontal position, for viewing sample and temperature simultaneously. Bright LCD digits are 6.4mm (1/4") high and easier to read than pointer or mercury thermometers.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a one-year warranty and 1.5V battery.
Customers Who Bought This Also Bought