To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:7
- Packaging:Box of 6 Pints
- Color:yellow
- Description:pH Buffer
- Size:475 ml
- pH Accuracy:±0.01
- Supplier No.:003822
Specifications
About this item
pH buffers for pH probe calibration.
Each buffer case contains 6 individual pints (plastic). Buffer cases are available with a single buffer type or with an assortment of buffers (2 pints of pH 4, 7, and 10 buffers).