Spectrophotometer Accessories, Thermo Scientific
Catalog #: 22365-003
Supplier: Thermo Scientific
Specifications
- Description:Test Tube Cuvette, Optical Glass; 3/4" Diameter (Path Length) and 150 mm Tall, Requires Light Shield (22358-008) and Test Tube Adapter (22355-000)
- Cat. no.:22365-003
- For Use With:SPECTRONIC® 20+/20D+ Spectrophotometers
- Supplier No.:331781-000