Specifications
- Volume:1000 µl
- Packed:Bulk
- Cat. no.:22234-086
- Tip Style:Non-Graduated
- For Use With:Most Research-Grade Pipettors
- Supplier No.:T-1000-C-L
Specifications
About this item
For applications requiring high accuracy and reproducibility.
Tips are constructed using diamond-polished molds to eliminate the cavities and occlusions that cause sample retention. Made of a unique polymer that resists DNA, RNA, and protein adherence, tips will not denature DNA. Manufactured without any secondary substances such as fillers, silicone, or heavy metals, which can contaminate samples.
- Ideal for transferring viscous DNA, RNA, and protein solutions
- Autoclavable under normal autoclaving guidelines
For applications requiring high accuracy and reproducibility.
Tips are constructed using diamond-polished molds to eliminate the cavities and occlusions that cause sample retention. Made of a unique polymer that resists DNA, RNA, and protein adherence, tips will not denature DNA. Manufactured without any secondary substances such as fillers, silicone, or heavy metals, which can contaminate samples.
Tips are constructed using diamond-polished molds to eliminate the cavities and occlusions that cause sample retention. Made of a unique polymer that resists DNA, RNA, and protein adherence, tips will not denature DNA. Manufactured without any secondary substances such as fillers, silicone, or heavy metals, which can contaminate samples.
Presterilized tips are exposed to e-beam radiation for protection from contamination.
All tips feature the 'Uni-grip' design, which assures an airtight seal with most major brands of pipettors.