Specifications
- Size:6×100 tablet
- Cat. no.:21834-109
- pH:6.5
- Supplier No.:1500
- Packing Type:Bottles
Specifications
About this item
Effervescent detergent tablet for lab pipette cleaning. Maximum cleaning in syphon pipette washers to clean pipettes and tubes. Slow release bubble action and free rinsing produce reliable results and no interfering residues.
- Concentrated
- Biodegradable and readily disposable
- Free rinsing to give you reliable results and no interfering residues
- Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
Special slow release tablet formulation for pipette cleaning. Maximum cleaning performance from your syphon pipette washers.
It can be used to clean pipettes and tubes with syphon pipette washers in hospitals, clinical, educational, R&D and industrial laboratories and cosmetics manufacturing.
And it can be used to remove soil, grit, grime, grease, fats, oils, blood, tissue, salt deposits, particulates, chemicals, solvents and radioisotopes.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.
Cleaning method: Syphon pipette and tube rinser.