VWR® Talon® Clamp Replacement Sleeves
Catalog #: 21572-900
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:Medium Two-Prong, Green Vinyl
- Cat. no.:21572-900
Specifications
About this item
Replacement sleeves are easily removed for cleaning or replacement.
- Vinyl or fiberglass fabrication
- Fiberglass sleeves suitable for high-temperature applications
- Easily removed for cleaning
Available in vinyl or fiberglass. Fiberglass sleeves are recommended for all applications above 100 °C (212 °F).
Customers Who Bought This Also Bought