VWR® Talon® Jumbo Holders
Catalog #: 21572-556
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:Clamp Holder
- Material:Aluminum
- Cat. no.:21572-556
Specifications
About this item
Holder secures clamps to lab frames and ring stands and accepts rods up to 21mm (13/16") in diameter.
- Electropolished stainless steel or aluminum construction
- Secure support of clamps
- Accepts rods of a range of diameters
Jumbo holders provide sturdy attachments of clamps to lab frames and ring stands.
Customers Who Bought This Also Bought