Accessories for TLC Developing Chamber, General Glass Blowing
Catalog #: 21432-794
Supplier: General Glass Blowing
Accessories for TLC Developing Chamber, General Glass Blowing
Catalog #: 21432-794
Supplier: General Glass Blowing
Specifications
- Description:Replacement Lid, Plain Glass
- Cat. no.:21432-794
- Supplier No.:80-4
Specifications
About this item
Customers Who Bought This Also Bought
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
411,94 $ to 1 303,38 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
150,16 $ to 1 502,77 $