Order Entry
ContactUsLinkComponent
80 PLC MIC TUBE RAC YEL PK5
80-Well Micro Tube Racks, Bio Plas
Catalog #: 21150-304
Supplier:  Bio Plas
80 PLC MIC TUBE RAC YEL PK5
80-Well Micro Tube Racks, Bio Plas
Catalog #: 21150-304
Supplier:  Bio Plas

Specifications

  • Color:
    Yellow
  • Cat. no.:
    21150-304
  • Supplier No.:
    0066

Specifications

About this item

High quality

  • Popular fraction collector plate-style rack
  • Easily fill and transfer 80 tubes in compact arrangement (5 x 16 array)
  • Rack dimensions are 9" L x 21/16" D, with a well diameter of 10.5 mm
  • Autoclavable

Each rack holds 1.5mL to 2.0mL microtubes in a 5x16 array. Tubes sit in large wells with conical bottoms. Each polypropylene rack is ideal for freezer storage.

Made in the USA.

Packaging: Racks are packaged 5 per pack, 4 packs per case. Assorted packages include one each: Blue, Green, Orange, Pink, and Yellow.
Customers Who Bought This Also Bought