VWR® PCR Tubes and Caps
Catalog #: 20170-010
Supplier: VWR International
Specifications
- Cap Type:Domed
- Capacity:0.2 ml
- Format:Tubes
- Color:Natural
- Environmentally Preferable:Sustainable Packaging,Low Manufacturing Impact
Specifications
About this item
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
- Autoclavable
- Chemically inert
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Tubes are available with bubble-top or flat-top caps, both of which fit precisely to ensure a positive, contamination-free environment.
Customers Who Bought This Also Bought