ContactUsLinkComponent
SCIENCEWARE SUPER GLUE
SP Bel-Art Super Glue, Bel-Art Products, a part of SP
Catalog #: 19807-000
SCIENCEWARE SUPER GLUE
SP Bel-Art Super Glue, Bel-Art Products, a part of SP
Catalog #: 19807-000
Specifications
  • Volume:
    3 g (0.11 fl. oz.)
  • Description:
    Super Glue
  • Cat. no.:
    19807-000
  • Supplier No.:
    240060000
Specifications
About this item
A drop of fast-setting Scienceware® Super Glue provides an amazingly strong repair or assembly joint.
  • Fast-setting
  • Solvent-free
  • Colorless and transparent when set
Use on metals, rubber, wood, most plastics, or semi-porous materials. Dissimilar materials may be joined as well. Scienceware® Super Glue is an ethyl-cyanoacrylate adhesive. It is solvent-free and requires no catalyst or heat. Colorless and transparent when set. Supplied 24 per case. No air shipment.
Customers Who Bought This Also Bought