BRAND Reagant Reservoir, BrandTech
Catalog #: 18900-320
Supplier: Brandtech
Specifications
- Capacity:60 ml
- Sterility:Non sterile
- Description:Reagent Reservoir with Lid
- Material:PP
- Supplier No.:703459
Specifications
About this item
This reagent reservoir is made from heavy-duty polypropylene to withstand autoclaving.
- Made of rugged, reusable polypropylene
- Autoclavable at 121 °C for 20 minutes
- Supplied with lid
A lid is included to help protect against sample contamination. Suitable for use with all multichannel pipettes.
Customers Who Bought This Also Bought