Single stand for Transferpette® multi-channel pipettes, Brand
Catalog #: 18900-124
Supplier: Brandtech
Specifications
- Description:Pipette Stand
- Cat. no.:18900-124
- Supplier No.:703440
Specifications
About this item
Single stand for all Transferpette® multi-channel Pipettes up to 300 µl.
Customers Who Bought This Also Bought