Laboratory Gas Lighters, G.C. Fuller
Catalog #: 18385-223
Supplier: G C Fuller
Specifications
- Description:Replacement flint tips
- Cat. no.:18385-223
- Supplier No.:3001X
Specifications
About this item
For lighting gas burners.
Designed with a rust-resistant zinc-plated spring handle and deep gas trap for quick ignitions. File rotates for fresh sparking surface. Each flint tip produces up to 2000 ignitions.
Customers Who Bought This Also Bought