Assay/Button Brush, Mill-Rose
Catalog #: 17210-008
Supplier: Mill-Rose
Specifications
- Bristle Material:Nylon
- Brush Head Diameter:16 mm (3/5")
- Total length:11.4 cm (5")
- Cat. no.:17210-008
- Supplier No.:MR-8888
Specifications
About this item
Stiff black nylon bristles for quick cleaning button samples for assays.
Flat-trimmed brushes have 0.014 gauge bristles mounted within a stainless steel tube. Double brush head.
Customers Who Bought This Also Bought