Order Entry
ContactUsLinkComponent
Buret and Pipet Brush
Buret and Pipet Brush, Mill-Rose
Catalog #: 17080-004
Supplier:  Mill-Rose
Buret and Pipet Brush
Buret and Pipet Brush, Mill-Rose
Catalog #: 17080-004
Supplier:  Mill-Rose

Specifications

  • Bristle Material:
    Nylon
  • Handle Material:
    Wire
  • Total length:
    43 cm (17")
  • Cat. no.:
    17080-004
  • Supplier No.:
    72115W

Specifications

About this item

Soft white nylon brush is specifically designed for cleaning pipets and burets.

  • Two different brush widths allow the simultaneous cleaning of delivery tubes and body
  • A flexible twisted wire handle with hanging loop secures the bristles
Customers Who Bought This Also Bought