Order Entry
ContactUsLinkComponent
KIMAX Cylindrical Weighing Bottles, Regular and Tall Form
KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Catalog #: 16713-127
KIMAX Cylindrical Weighing Bottles, Regular and Tall Form
KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Catalog #: 16713-127

Specifications

  • Capacity:
    30 ml (1 oz.)
  • Dimensions:
    30 I.D. x 60 H, mm
  • ST Joint No.:
    34/12
  • Cat. no.:
    16713-127
  • Supplier No.:
    15146-3060

Specifications

About this item

With airtight, cap-style glass [ST] stopper.

  • Area for marking on each bottle
  • Capacity is to base of neck

Designed from Federal Specification NNN-1493, Type I, Style II requirements.

Customers Who Bought This Also Bought