Nalgene® Rectangular Carboys with Spigot and Handle, High-Density Polyethylene, Thermo Scientific
Catalog #: 16338-039
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:20 L (5.3 gal.)
- Dimensions:31.8 x 21.9 x 38.1 cm (121/2 x 85/8 x 15")
- Cat. no.:16338-039
- Supplier No.:2320-0050
Specifications
About this item
Excellent for storage and dispensing of buffers, acids, and bases.
- Tough, rigid translucent carboy features molded-in graduations
- Wide mouth allows easy, efficient cleaning
- Built-in shoulder loops have sturdy handle attached
- Leakproof
Excellent for storage and dispensing of buffers, acids, and bases.
Ordering information: Supplied with carboy, polypropylene spigot, and 100-415 screw cap. For replacement caps, see cap 16219-106 in 16219-030 series.
Customers Who Bought This Also Bought