Nalgene® Carboys with Spigot and Handles, Low-Density Polyethylene, Thermo Scientific
Catalog #: 16334-220
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:83B
- Capacity:20 L (5.3 gal.)
- Cat. no.:16334-220
- Supplier No.:2318-0050
Specifications
About this item
Suitable for collecting and dispensing distilled water, reagents, and acids.
- Leakproof
Spigots accept 16 mm (⁵/₈") Int.Ø tubing, and include an HDPE tubing adapter that takes tubing with 6 to 8 mm (¼ to ⁵/₁₆") Int.Ø. The ETFE spigot is for use with applications with selected solvents and chemicals in which polypropylene spigots are unsatisfactory.
Graduated to deliver in 1 gal. and 5 L increments. Spigot boss molded in one piece with bottle, not welded on, for strength and leak resistance. Spigot comes with adapter to accommodate a variety of tubing sizes. The 3.8 L (1 gal.) size does not have handles.
Ordering information: Supplied with carboy, polypropylene spigot, and screw cap. For replacement caps, see 16219-122 series.
Customers Who Bought This Also Bought