Nalgene® Cryoware Marker Set, Thermo Scientific
Catalog #: 16211-130
Supplier: Thermo Fisher Scientific
Specifications
- Color:Black
- Description:Marker
- Cat. no.:16211-130
- Supplier No.:6313-0020
Specifications
About this item
Permanent markers with extra-fine tips for marking vials, cardboard, and plastic boxes
Excellent for permanent, colorful identification of cryogenic samples. Ink will not fade at ultra-low temperatures. Markers are not alcohol resistant.
Customers Who Bought This Also Bought