Order Entry
ContactUsLinkComponent
Round Bottles, Polypropylene, Wide Mouth
Round Bottles, Polypropylene, Wide Mouth, Taral
Catalog #: 16196-080
Round Bottles, Polypropylene, Wide Mouth
Round Bottles, Polypropylene, Wide Mouth, Taral
Catalog #: 16196-080

Specifications

  • Cap Size:
    89 mm
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    16196-080
  • Supplier No.:
    16-89-NPPWC

Specifications

About this item

These handy, unbreakable bottles are suitable for use with chemicals.

  • Bottles are autoclavable and caps are not.

Included white caps have polyvinyl liners. Packaged in polyethylene bags.

Customers Who Bought This Also Bought