Aluminum Seals, Tear-Off, WHEATON®
Catalog #: 16171-840
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Material:Standard Aluminum
- Cat. no.:16171-840
- For O.D.:13 mm
- Supplier No.:224192-01
Specifications
About this item
Seals can be punctured by syringe or completely removed and contents poured from bottle
Shelf packs of 100.
Customers Who Bought This Also Bought