WHEATON® Serum Vials, Borosilicate Glass, DWK Life Sciences
Catalog #: 16171-286
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap included:No
- Capacity:5.00 ml
- Color:Clear
- Cat. no.:16171-286
- Diameter ×H:22×40 mm
- Supplier No.:223685
Specifications
About this item
Tubular design provides excellent clarity and dimensional consistency from vial to vial.
- Autoclavable
Clear vials manufactured from low extractable borosilicate glass.Amber glass protect light-sensitive samples.
Specially designed bottom radius adds strength for lyophilization applications.
Certifications: Amber glass conforms to USP Type I requirements.Borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Packaging: Vials are packaged in shrink-wrapped modules reduce particulate contamination.
Customers Who Bought This Also Bought