Nalgene® PassPort™ Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Catalog #: 16129-756
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:63-415
- Capacity:1 L (33.8 oz.)
- Cat. no.:16129-756
- Supplier No.:2199-0032
Specifications
About this item
Safe, durable, cost-efficient bottles for packaging and transporting hazardous materials.
- Molded into bottom: IP2 marking, material, volume, and registration notch
- Heavy-duty walls
- Leakproof
Evaluated at 15 psi (103 kPa) per 49 CFR 173.27 (c) (2), ICAO Technical Instructions Part 3-1; 1.1.6.1, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Recommended for customers who are designing, assembling, and certifying their own combination packaging. Split- and puncture-resistant. Wide mouth allows for easy filling of dry materials or liquids.
Caution: Warning: Do not use under pressure or vacuum. The application of pressure or vacuum to products not designed for such use may result in product failure, damage to property, or personal injury.
Customers Who Bought This Also Bought