Order Entry
ContactUsLinkComponent
NALGENE* Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth
Nalgene® Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Catalog #: 16129-706
NALGENE* Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth
Nalgene® Bottles and Carboys, Fluorinated High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Catalog #: 16129-706

Specifications

  • Cap Size:
    38-430
  • Capacity:
    2 L (67.6 oz.)
  • Cat. no.:
    16129-706
  • Supplier No.:
    2097-0005

Specifications

About this item

Safe, durable, cost-efficient line of narrow mouth bottles and carboys feature fluorinated surfaces (inside and outside), for improved barrier properties and reduced solvent absorption and penetration.

  • Split- and puncture-resistant
  • With polypropylene screw closure
  • Heavy-duty walls, Leakproof
  • 10 and 20 L sizes feature molded-in graduations

Fluorination enhances long-term container performance and prevents or reduces permeation loss. Useful with most aggressive organic solvents.

Ordering information: Closures not available separately.
Customers Who Bought This Also Bought